Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 25 - 26 | 3510 | 79 | |
| XM_003641367.4 | XP_003641415.1 | 27 - 28 | 79 | |||
| XM_015286526.2 | XP_015142012.1 | 27 - 28 | 79 | |||
| XM_015286527.2 | XP_015142013.1 | 28 - 29 | 79 | |||
| XM_015286528.2 | XP_015142014.1 | 23 - 24 | 79 | |||
| GenBank mRNA | AB046524.1 | - | 25 - 26 | 3510 | 79 | |
More Information
Set Membership: |
|

OVOA
Genomic Map