Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 31 - 32 | 4301 | 85 | |
| XM_003641367.4 | XP_003641415.1 | 33 - 34 | 5065 | 85 | ||
| XM_015286526.2 | XP_015142012.1 | 33 - 34 | 5291 | 85 | ||
| XM_015286527.2 | XP_015142013.1 | 34 - 35 | 5245 | 85 | ||
| XM_015286528.2 | XP_015142014.1 | 29 - 30 | 4631 | 85 | ||
| GenBank mRNA | AB046524.1 | - | 31 - 32 | 4301 | 85 | |
More Information
Set Membership: |
|

OVOA
Genomic Map