Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 42 - 43 | 5780 | 63 | |
| XM_003641367.4 | XP_003641415.1 | 44 - 45 | 6544 | 63 | ||
| XM_015286526.2 | XP_015142012.1 | 44 - 45 | 6770 | 63 | ||
| XM_015286527.2 | XP_015142013.1 | 45 - 46 | 6724 | 63 | ||
| XM_015286528.2 | XP_015142014.1 | 40 - 41 | 6110 | 63 | ||
| GenBank mRNA | AB046524.1 | - | 42 - 43 | 5780 | 63 | |
More Information
Set Membership: |
|

OVOA
Genomic Map