Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 45 - 46 | 6048 | 74 | |
| XM_003641367.4 | XP_003641415.1 | 47 - 48 | 6812 | 74 | ||
| XM_015286526.2 | XP_015142012.1 | 47 - 48 | 7038 | 74 | ||
| XM_015286527.2 | XP_015142013.1 | 48 - 49 | 6992 | 74 | ||
| XM_015286528.2 | XP_015142014.1 | 43 - 44 | 6378 | 74 | ||
| GenBank mRNA | AB046524.1 | - | 45 - 46 | 6048 | 74 | |
More Information
Set Membership: |
|

OVOA
Genomic Map