Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 7 - 8 | 1046 | 90 | |
| XM_003641367.4 | XP_003641415.1 | 9 - 10 | 1810 | 90 | ||
| XM_015286526.2 | XP_015142012.1 | 9 - 10 | 2036 | 90 | ||
| XM_015286527.2 | XP_015142013.1 | 10 - 11 | 1990 | 90 | ||
| XM_015286528.2 | XP_015142014.1 | 5 - 6 | 1376 | 90 | ||
| GenBank mRNA | AB046524.1 | - | 7 - 8 | 1046 | 90 | |
More Information
Set Membership: |
|

OVOA
Genomic Map