Assay Details
| Gene Symbol: C4BPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204664.2 | NP_989995.1 | 264 | 81 | ||
| XM_015298885.2 | XP_015154371.1 | 300 | 81 | |||
| XM_015298886.2 | XP_015154372.1 | 300 | 81 | |||
| XM_015298887.2 | XP_015154373.1 | 300 | 81 | |||
| GenBank mRNA | AB035592.1 | - | 265 | 81 | ||
| CR382436.1 | - | 287 | 81 | |||

C4BPM
Genomic Map