Assay Details
| Gene Symbol: C4BPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204664.2 | NP_989995.1 | 1312 | 100 | ||
| XM_015298887.2 | XP_015154373.1 | 1321 | 100 | |||
| GenBank mRNA | AB035592.1 | - | 1313 | 100 | ||
| CR382436.1 | - | 1336 | 100 | |||
More Information
Set Membership: |
|

C4BPM
Genomic Map