Assay Details
| Gene Symbol: CD40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204665.2 | NP_989996.1 | 5 - 6 | 539 | 96 | |
| GenBank mRNA | AJ293700.1 | - | 5 - 6 | 539 | 96 | |
| EF554721.1 | - | 5 - 6 | 517 | 96 | ||
More Information
Set Membership: |
|

CD40
Genomic Map