Assay Details
| Gene Symbol: PDGFC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204721.2 | NP_990052.1 | 3 - 4 | 591 | 71 | |
| XM_025149500.1 | XP_025005268.1 | 3 - 4 | 490 | 71 | ||
| GenBank mRNA | AB033829.1 | - | 3 - 4 | 593 | 71 | |
More Information
Set Membership: |
|

PDGFC
Genomic Map