Assay Details
| Gene Symbol: UCHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204825.1 | NP_990156.1 | 7 - 8 | 578 | 70 | |
| XM_015275276.2 | XP_015130762.1 | 7 - 8 | 814 | 70 | ||
| XM_025142482.1 | XP_024998250.1 | 7 - 8 | 640 | 70 | ||
| GenBank mRNA | AF167995.1 | - | 7 - 8 | 578 | 70 | |
| BX934698.1 | - | 8 - 9 | 694 | 70 | ||
More Information
Set Membership: |
|

UCHL3
Genomic Map