Assay Details
| Gene Symbol: RBBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204852.1 | NP_990183.1 | 2 - 3 | 238 | 73 | |
| XM_015297601.2 | XP_015153087.1 | 2 - 3 | 274 | 73 | ||
| GenBank mRNA | AF097750.1 | - | 2 - 3 | 238 | 73 | |
| CR390284.1 | - | 2 - 3 | 269 | 73 | ||
More Information
Set Membership: |
|

RBBP4
Genomic Map