Assay Details
| Gene Symbol: ZP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204879.1 | NP_990210.1 | 6 - 7 | 872 | 74 | |
| XM_025151486.1 | XP_025007254.1 | 6 - 7 | 2066 | 74 | ||
| GenBank mRNA | AB025428.1 | - | 6 - 7 | 872 | 74 | |
More Information
Set Membership: |
|

ZP4
Genomic Map