Assay Details
| Gene Symbol: CLU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204900.1 | NP_990231.1 | 951 | 88 | ||
| GenBank mRNA | AF119370.1 | - | 951 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLU| Gene Symbol: CLU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204900.1 | NP_990231.1 | 951 | 88 | ||
| GenBank mRNA | AF119370.1 | - | 951 | 88 | ||
Set Membership: |
|