Assay Details
| Gene Symbol: RHOB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204909.1 | NP_990240.1 | 1 | 852 | 64 | |
| GenBank mRNA | AF098515.1 | - | 1 | 852 | 64 | |
| U79758.1 | - | 1 | 606 | 64 | ||
More Information
Set Membership: |
|

RHOB
Genomic Map