Assay Details
Gene Symbol: TRHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204930.1 | NP_990261.1 | 1 - 2 | 793 | 123 | |
GenBank mRNA | Y18244.1 | - | 793 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
