Assay Details
Gene Symbol: ST3GAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205217.1 | NP_990548.1 | 5 - 6 | 983 | 73 | |
XM_015283081.2 | XP_015138567.1 | 6 - 7 | 1393 | 73 | ||
GenBank mRNA | JX035875.1 | - | 5 - 6 | 879 | 73 | |
X80503.1 | - | 983 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
