Assay Details
Gene Symbol: ST6GALNAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205233.1 | NP_990564.1 | 4 - 5 | 843 | 69 | |
GenBank mRNA | X77775.1 | - | 843 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
