Assay Details
| Gene Symbol: YAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205243.1 | NP_990574.1 | 6 - 7 | 1170 | 69 | |
| GenBank mRNA | X76483.1 | - | 1170 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

YAP1| Gene Symbol: YAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205243.1 | NP_990574.1 | 6 - 7 | 1170 | 69 | |
| GenBank mRNA | X76483.1 | - | 1170 | 69 | ||
Set Membership: |
|