Assay Details
| Gene Symbol: CSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205248.1 | NP_990579.1 | 178 | 63 | ||
| XM_015298693.2 | XP_015154179.1 | 129 | 63 | |||
| XM_025143676.1 | XP_024999444.1 | 384 | 63 | |||
| XM_025143677.1 | XP_024999445.1 | 129 | 63 | |||
| XM_025143678.1 | XP_024999446.1 | 129 | 63 | |||
| GenBank mRNA | X73831.1 | - | 178 | 63 | ||
More Information
Set Membership: |
|

CSRP1
Genomic Map