Assay Details
Gene Symbol: ANXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205351.1 | NP_990682.1 | 3 - 4 | 195 | 72 | |
XM_015278763.2 | XP_015134249.1 | 5 - 6 | 417 | 72 | ||
XM_025153799.1 | XP_025009567.1 | 7 - 8 | 3167 | 72 | ||
XM_025153800.1 | XP_025009568.1 | 6 - 7 | 3764 | 72 | ||
XM_025153801.1 | XP_025009569.1 | 5 - 6 | 399 | 72 | ||
XM_025153802.1 | XP_025009570.1 | 7 - 8 | 3164 | 72 | ||
XM_025153803.1 | XP_025009571.1 | 3 - 4 | 365 | 72 | ||
GenBank mRNA | KU749995.1 | - | 2 - 3 | 148 | 72 | |
KU749996.1 | - | 2 - 3 | 148 | 72 | ||
KU749997.1 | - | 2 - 3 | 148 | 72 | ||
KU749998.1 | - | 2 - 3 | 148 | 72 | ||
KU749999.1 | - | 2 - 3 | 148 | 72 | ||
KU750000.1 | - | 2 - 3 | 148 | 72 | ||
KU750001.1 | - | 2 - 3 | 148 | 72 | ||
KU750002.1 | - | 2 - 3 | 148 | 72 | ||
KU750003.1 | - | 2 - 3 | 148 | 72 | ||
KU750004.1 | - | 2 - 3 | 148 | 72 | ||
KU750005.1 | - | 2 - 3 | 148 | 72 | ||
KU750006.1 | - | 2 - 3 | 148 | 72 | ||
KU750007.1 | - | 2 - 3 | 148 | 72 | ||
KU750008.1 | - | 2 - 3 | 148 | 72 | ||
KU750009.1 | - | 2 - 3 | 148 | 72 | ||
KU750010.1 | - | 2 - 3 | 148 | 72 | ||
KU750011.1 | - | 2 - 3 | 148 | 72 | ||
KU750012.1 | - | 2 - 3 | 148 | 72 | ||
X53334.1 | - | 195 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
