Assay Details
| Gene Symbol: EEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205368.1 | NP_990699.1 | 12 - 13 | 2096 | 59 | |
| GenBank mRNA | U46663.1 | - | 12 - 13 | 2096 | 59 | |
More Information
Set Membership: |
|

EEF2
Genomic Map