Assay Details
| Gene Symbol: PRSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205384.1 | NP_990715.1 | 2 - 3 | 226 | 120 | |
| GenBank mRNA | CR390911.1 | - | 2 - 3 | 227 | 120 | |
| U15157.1 | - | 2 - 3 | 226 | 120 | ||
| Gene Symbol: LOC107054567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294158.2 | XP_015149644.1 | 2 - 3 | 323 | 120 | |
| Gene Symbol: LOC107054568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294165.2 | XP_015149651.1 | 2 - 3 | 241 | 120 | |
More Information
Set Membership: |
|

PRSS2
Genomic Map