Assay Details
Gene Symbol: KCNJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205404.1 | NP_990735.1 | 1 - 2 | 717 | 77 | |
GenBank mRNA | L35555.1 | - | 1 - 2 | 717 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
