Assay Details
| Gene Symbol: EXFABP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205422.1 | NP_990753.1 | 5 - 6 | 526 | 88 | |
| GenBank mRNA | M25784.1 | - | 5 - 6 | 526 | 88 | |
| X61199.1 | - | 266 | 88 | |||
More Information
Set Membership: |
|

EXFABP
Genomic Map