Assay Details
Gene Symbol: LECT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205478.2 | NP_990809.2 | 4 - 5 | 531 | 87 | |
GenBank mRNA | CR388911.1 | - | 4 - 5 | 518 | 87 | |
M29449.1 | - | 4 - 5 | 531 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
