Assay Details
| Gene Symbol: CNTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205492.1 | NP_990823.1 | 1 - 2 | 207 | 73 | |
| XM_015286770.2 | XP_015142256.1 | 2 - 3 | 423 | 73 | ||
| GenBank mRNA | M80827.1 | - | 1 - 2 | 207 | 73 | |
More Information
Set Membership: |
|

CNTF
Genomic Map