Assay Details
Gene Symbol: CST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205500.2 | NP_990831.2 | 2 - 3 | 396 | 122 | |
GenBank mRNA | AJ395644.1 | - | 2 - 3 | 338 | 122 | |
BU119189.1 | - | 2 - 3 | 331 | 122 | ||
BU140104.1 | - | 2 - 3 | 334 | 122 | ||
CN210879.1 | - | 2 - 3 | 385 | 122 | ||
J05077.1 | - | 2 - 3 | 394 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
