Assay Details
| Gene Symbol: RNF114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001767.2 | NP_001001767.2 | 3 - 4 | 400 | 77 | |
| GenBank mRNA | AJ719560.1 | - | 3 - 4 | 400 | 77 | |
| AY604724.1 | - | 3 - 4 | 427 | 77 | ||
More Information
Set Membership: |
|

RNF114
Genomic Map