Assay Details
Gene Symbol: CMTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001778.2 | NP_001001778.2 | 1 - 2 | 146 | 93 | |
GenBank mRNA | AB106870.1 | - | 1 - 2 | 144 | 93 | |
AJ720911.1 | - | 1 - 2 | 146 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
