Assay Details
| Gene Symbol: BIRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007822.1 | NP_001007823.1 | 6 - 7 | 2602 | 70 | |
| GenBank mRNA | AF008592.1 | - | 5 - 6 | 2477 | 70 | |
| CR407264.1 | - | 5 - 6 | 681 | 70 | ||
| U27466.1 | - | 5 - 6 | 1854 | 70 | ||
More Information
Set Membership: |
|

BIRC3
Genomic Map