Assay Details
| Gene Symbol: APOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011692.1 | NP_001011692.1 | 3 - 4 | 321 | 80 | |
| GenBank mRNA | AY735998.1 | - | 3 - 4 | 321 | 80 | |
| BX934317.2 | - | 3 - 4 | 322 | 80 | ||
More Information
Set Membership: |
|

APOD
Genomic Map