Assay Details
| Gene Symbol: EPGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012404.1 | NP_001012404.1 | 2 - 3 | 718 | 68 | |
| XM_015276322.2 | XP_015131808.1 | 2 - 3 | 3524 | 68 | ||
| GenBank mRNA | AY885230.1 | - | 2 - 3 | 718 | 68 | |
More Information
Set Membership: |
|

EPGN
Genomic Map