Assay Details
| Gene Symbol: EDARADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012405.2 | NP_001012405.2 | 5 - 6 | 298 | 81 | |
| GenBank mRNA | AJ720393.1 | - | 5 - 6 | 298 | 81 | |
More Information
Set Membership: |
|

EDARADD
Genomic Map