Assay Details
| Gene Symbol: EIF3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012535.2 | NP_001012553.1 | 12 - 13 | 1686 | 61 | |
| GenBank mRNA | AJ851472.1 | - | 12 - 13 | 1687 | 61 | |
More Information
Set Membership: |
|

EIF3L
Genomic Map