Assay Details
Gene Symbol: NUP50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012541.1 | NP_001012559.1 | 2 - 3 | 319 | 65 | |
GenBank mRNA | AJ851427.1 | - | 2 - 3 | 319 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
