Assay Details
Gene Symbol: C26H6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012548.1 | NP_001012566.1 | 813 | 74 | ||
XM_015298899.2 | XP_015154385.1 | 989 | 74 | |||
XM_015298900.2 | XP_015154386.1 | 816 | 74 | |||
XM_015298901.2 | XP_015154387.1 | 2453 | 74 | |||
GenBank mRNA | AJ851806.1 | - | 813 | 74 |
