Assay Details
| Gene Symbol: C26H6orf89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012548.1 | NP_001012566.1 | 942 | 68 | ||
| XM_015298899.2 | XP_015154385.1 | 1118 | 68 | |||
| XM_015298900.2 | XP_015154386.1 | 945 | 68 | |||
| XM_015298901.2 | XP_015154387.1 | 2582 | 68 | |||
| GenBank mRNA | AJ851806.1 | - | 942 | 68 | ||
More Information
Set Membership: |
|

C26H6orf89
Genomic Map