Assay Details
Gene Symbol: NRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012549.1 | NP_001012567.1 | 715 | 62 | ||
GenBank mRNA | AJ851798.1 | - | 715 | 62 | ||
HQ439435.1 | - | 680 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
