Assay Details
| Gene Symbol: CTDSPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012772.1 | NP_001012790.1 | 2 - 3 | 487 | 80 | |
| XM_015292114.2 | XP_015147600.1 | 3 - 4 | 480 | 80 | ||
| XM_015292115.2 | XP_015147601.1 | 3 - 4 | 861 | 80 | ||
| XM_025153866.1 | XP_025009634.1 | 3 - 4 | 485 | 80 | ||
| GenBank mRNA | AJ851503.1 | - | 2 - 3 | 487 | 80 | |
| BX930341.1 | - | 3 - 4 | 510 | 80 | ||
More Information
Set Membership: |
|

CTDSPL2
Genomic Map