Assay Details
| Gene Symbol: FBXL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012786.1 | NP_001012804.1 | 3 - 4 | 1813 | 66 | |
| GenBank mRNA | AJ851502.1 | - | 3 - 4 | 1813 | 66 | |
More Information
Set Membership: |
|

FBXL18
Genomic Map