Assay Details
| Gene Symbol: UBE2N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012810.1 | NP_001012828.1 | 2 - 3 | 354 | 83 | |
| GenBank mRNA | AJ851495.1 | - | 2 - 3 | 354 | 83 | |
| CR387699.1 | - | 2 - 3 | 324 | 83 | ||
More Information
Set Membership: |
|

UBE2N
Genomic Map