Assay Details
Gene Symbol: TMEM164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012886.1 | NP_001012904.1 | 5 - 6 | 882 | 67 | |
XM_015278293.1 | XP_015133779.1 | 4 - 5 | 217 | 67 | ||
GenBank mRNA | AJ721046.1 | - | 5 - 6 | 882 | 67 | |
BX931185.2 | - | 4 - 5 | 277 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
