Assay Details
| Gene Symbol: SUCLG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012892.1 | NP_001012910.1 | 2 - 3 | 206 | 74 | |
| XM_025149734.1 | XP_025005502.1 | 2 - 3 | 327 | 74 | ||
| GenBank mRNA | AJ719851.1 | - | 2 - 3 | 206 | 74 | |
More Information
Set Membership: |
|

SUCLG1
Genomic Map