Assay Details
| Gene Symbol: HACD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012920.4 | NP_001012938.2 | 5 - 6 | 537 | 91 | |
| GenBank mRNA | AJ720797.1 | - | 6 - 7 | 679 | 91 | |
| BU356427.1 | - | 5 - 6 | 486 | 91 | ||
| CR352863.1 | - | 5 - 6 | 478 | 91 | ||
More Information
Set Membership: |
|

HACD2
Genomic Map