Assay Details
Gene Symbol: DNAJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012945.1 | NP_001012963.1 | 2 - 3 | 415 | 56 | |
GenBank mRNA | AJ720331.1 | - | 2 - 3 | 415 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
