Assay Details
| Gene Symbol: WISP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024579.1 | NP_001019750.1 | 1 - 2 | 100 | 115 | |
| XM_015283082.2 | XP_015138568.1 | 1 - 2 | 167 | 115 | ||
| GenBank mRNA | BX933985.2 | - | 1 - 2 | 133 | 115 | |
| BX934666.1 | - | 1 - 2 | 157 | 115 | ||
| DQ003338.1 | - | 1 - 2 | 100 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015283082.2 transcripts.
Set Membership: |
|

WISP1
Genomic Map