Assay Details
| Gene Symbol: CD1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024582.1 | NP_001019753.1 | 4 - 5 | 913 | 103 | |
| GenBank mRNA | AY849320.1 | - | 4 - 5 | 913 | 103 | |
More Information
Set Membership: |
|

CD1B
Genomic Map