Assay Details
Gene Symbol: TLR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024586.1 | NP_001019757.1 | 1 | 2454 | 148 | |
XM_015283741.2 | XP_015139227.1 | 3 | 6316 | 148 | ||
XM_015283742.2 | XP_015139228.1 | 2 | 3018 | 148 | ||
XM_025148815.1 | XP_025004583.1 | 3 | 9040 | 148 | ||
GenBank mRNA | DQ004555.1 | - | 1 | 2454 | 148 | |
JF767211.1 | - | 1 | 2454 | 148 | ||
JF767212.1 | - | 1 | 2454 | 148 | ||
JF767214.1 | - | 1 | 2454 | 148 | ||
JF767216.1 | - | 1 | 2454 | 148 | ||
JF767217.1 | - | 1 | 2454 | 148 | ||
JF767219.1 | - | 1 | 2454 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
