Assay Details
Gene Symbol: SSTR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024834.1 | NP_001020005.1 | 1 | 953 | 127 | |
XM_015294245.2 | XP_015149731.1 | 2 | 1209 | 127 | ||
XM_015294246.2 | XP_015149732.1 | 2 | 1486 | 127 | ||
GenBank mRNA | AY954512.1 | - | 1 | 953 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
