Assay Details
| Gene Symbol: RHOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029849.1 | NP_001025020.1 | 1 | 307 | 113 | |
| GenBank mRNA | AF098514.1 | - | 1 | 307 | 113 | |
| U79759.1 | - | 224 | 113 | |||
More Information
Set Membership: |
|

RHOC
Genomic Map